Corrugated Mailers, Associated Bag
Catalogue # 76250-324
Fournisseur: Associated Bag
Spécifications
- Color:White
- Description:Corrugated mailers, 200 lbs. test/32 lbs. ect
- Dimensions:8×5×4"
- Cat. no.:76250-324
Spécifications
À propos de cet article
Reusable mailers protect contents during storage and shipping.
- Requires no glue, tape, or staples