Order Entry
Canada
Orders LinkContactUsLinkComponent
Connector Plug Stereo, Digi-Key

Connector Plug Stereo, Digi-Key

Six Position Receptacle Connector
Catalog # 76253-848
Supplier #:  5-535541-4
Connector Plug Stereo, Digi-Key
Catalog # 76253-848
Connector Plug Stereo, Digi-Key

Six Position Receptacle Connector

Price
$14.62
Log in for availability
Each
Product Details & Documents

Connector Plug; Board to Board or Cable.

  • Length: 3.18 mm
  • Material: Polyester, Glass Filled
  • Size: 2.54 mm (0.100")
Specifications
  • Description:
    Six Position Receptacle Connector
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.