Cable Stripper, Duratool G312, Newark
Catalog # 76248-654
Supplier: NEWARK/ELEMENT 14 TE
Specifications
- Description:Duratool Coaxial Cable Stripper
- Cat. no.:76248-654
Specifications
About this item
Cable stripping tool for coaxial cables, rotary, adjustable (three adjustable blade modules).