START Replacement Blade for Tape Dispenser, Jensen Tools
Catalog # 76252-196
Supplier: TESTEQUITY LLC TE
Specifications
- Description:Replacement Blade Set
- Cat. no.:76252-196
Specifications
About this item
Replacement blade set for ZCM1000 series of tape dispenser.
- Constructed of hard-coated stainless steel
Blade cuts most standard tape for up to 200000 cuts (dependent upon tape material and adhesive type).
Delivery: Blade set includes both upper and lower blades.