ContactUsLinkComponent
ESD Badge Holders with Zipper, Menda
ESD Badge Holders with Zipper, Menda
  76251-160
 :  Menda
ESD Badge Holders with Zipper, Menda
  76251-160
 :  Menda
 :  35017
  • Description:
    Horizontal ESD Card Holder
  • Dimensions:
    35/8×23/8"
  • Cat. no.:
    76251-160
ESD Badge holders with zipper closure protects badge from moisture and dirt as well as ensures contents stay inside.
  • Tribocharging less than 100 V
  • Made with custom dissipative material
  • Available in vertical and horizontal shapes
Ideal for use by electronic manufacturers working with static sensitive components. Highly visible with yellow header and ESD symbol allowing people to immediately know the ESD control properties: low charging, antistatic and dissipative.
 : Includes inner anti-print, non-ESD vinyl liner which will prevent ink transfer from document to the document holder.