Tacki-Pak™ Component Box, Conductive Containers
76248-308
- Description:Black
- Dimensions:3.5×4.625×0.5"
- Cat. no.:76248-308
Tacki-Pak™ provides an ESD-Safe, non-adhesive, tacky surface that will limit component movement. No longer will parts slide into each other causing damage.
- Tack level I rating range 2.0 - 4.0 (rating is in foot pounds of force)
- Box surface resistance 103- 106 ohms
- Tacki surface resistance 105 - 109 ohms
- Reusable - clean with IPA (wipes) to remove FOD and rejuvenate surface tack level
This is a flat, user friendly surface that minimizes residue transfer to the component. It has stable, reusable and cleanable ESD-Safe properties.