Burgundy CCH Adjustable ESD Wrist Strap Set, Botron
Catalog # 76267-634
Supplier: BOTRON COMPANY INC.
Specifications
- Color:Burgundy band/Black coil cord
- Material:Knitted 3/4" wide Nylon
- Resistor:1 MΩ
- Cat. no.:76267-634
- Cord Length:6'
Specifications
About this item
Constant contact hinge adjustable wrist strap set is available with a standard coil cord (PVC) design. They provide the performance and reliability of a metal band with the comfort of cloth.
- The blue knitted material has a soft outer area and a 5/8" conductive area on the inside
- The constant contact hinge ensures continuity, reducing latent failures
- Back pate of stainless steel and face plate of composite plastic
Anti static wrist strap sets should be securely attached either directly to the ground or via banana jack to an ESD table mat or ESD floor mat. Compatible with all constant monitors, table and floor mat grounds, as well as ESD bench grounds.
To install open the face of the wrist strap, sliding it over hand onto wrist, pull end of wrist strap tightly, close face of strap, attach coil cord, plug into continuous monitor or other recommended grounding devices.