Orders LinkContactUsLinkComponent
Adapters, Digi-Key
Adapter Connector, 0.126" (3.20 mm) I-Ø, 0.327" (8.30 mm) O-Ø
  76250-608
 :  BPJJ02AUX
Adapters, Digi-Key
  76250-608
Adapters, Digi-Key
Adapter Connector, 0.126" (3.20 mm) I-Ø, 0.327" (8.30 mm) O-Ø
$41.28
Each
Adapter connector Phono (RCA) jack to Phono (RCA) jack panel mount.
  • Brass body with gold finish
  • Nickel plated contact material
  • Red insulation
  • Unshielded
Female to female connector.
 : Includes 1 connector, 1 nut and 1 washer.
  • Description:
    Adapter Connector, 0.126" (3.20 mm) I-Ø, 0.327" (8.30 mm) O-Ø