About this item
The new XSR analytical balances simplify weighing processes and provide years of accurate results.
- Results protocol
- Storage of methods
- Balance ready StatusLight
- LevelControl: level warning with graphical levelling guide
- Ionizer module ready, LabX ready and calibry ready
The XSR analytical balances, deliver high weighing accuracy and provide convenient features that allow the operator to fulfil weighing workflows seamlessly. Features, such as SmartGrid weighing pan, automated doors, and the results protocol, simplify routine weighing tasks and take the stresses and strains out of weighing.
Built-in methods support your everday weighing tasks. Develop and save your own weighing method specific to your needs and start it directly from the method window. StatusLight™ checks that all the conditions for correct weighing have been met to ensure your results are reliable.
Features with 7" color touchscreen (glove compatible), ergoClip for direct dosing, SmartSens for touch-less door operation, SmartTrac dosing guide automatic draft shield doors, proFACT advanced internal adjustment, MinWeigh warning function and SmartGrid weighing pan.
It has seamless process, tolerance profiles, efficient operations, adjustment and routine test history, change history function, user management and Active Temperature Control system (ATC™).
Specifications
- Catalog No:
- 76292-096
- 76292-102
- 76292-104
- 76292-098
- 76292-100
- 76292-158
- 76292-106
- 76292-108
- 76292-094
- Communication interface:
- 4×USB, 1×LAN, RS232 Compatible
- 4×USB, 1×LAN, RS232 Compatible
- 4×USB, 1×LAN, RS232 Compatible
- 4×USB, 1×LAN, RS232 Compatible
- 4×USB, 1×LAN, RS232 Compatible
- 4×USB, 1×LAN, RS232 Compatible
- 4×USB, 1×LAN, RS232 Compatible
- 4×USB, 1×LAN, RS232 Compatible
- 4×USB, 1×LAN, RS232 Compatible