Order Entry
Canada
ContactUsLinkComponent
Copper(II) bromide 50% in aqueous solution, AR®, Macron Fine Chemicals™
Copper(II) bromide 50% in aqueous solution, AR®, Macron Fine Chemicals™
Catalog # CAMK4895-22
CAS Number:  
undefined
Copper(II) bromide 50% in aqueous solution, AR®, Macron Fine Chemicals™
Catalog # CAMK4895-22
Supplier Number:  MACR4895-22
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    HDPE poly drum
  • Size:
    50 lbs.
  • Cat. no.:
    CAMK4895-22

Specifications

Specification Test Results

  • Appearance (dark brown/black solution):
    Passes Test
  • Assay (CuBr₂):
    49 - 51 %
  • Insoluble Matter:
    ≤0.005 %
  • Sulfate (SO₄):
    ≤0.005 %
  • Chloride (Cl):
    ≤0.15 %
  • Substances not Precipitated by H₂S (as SO₄):
    ≤0.10 %
  • Iron (Fe):
    ≤0.003 %
  • pH of 10% solution:
    2.7 - 3.0

Specification Test Results

About this item

 :
 :223.35 g/mol
Density:1.50 kg (1 L)
ADR:8,II
 :Management of Change (MOC) category = N
UN:3264
 :GHS07
 :GHS05
 :GHS09
CAS Number:7789-45-9
Formula:Br₂Cu
MDL Number:MFCD00010970