ContactUsLinkComponent
Tefzel® Flangeless Tubing Nuts, Chemglass
Tefzel® Flangeless Tubing Nuts, Chemglass
Catalog # 80060-480
Supplier:  Chemglass
Small Business Enterprise
undefined
Tefzel® Flangeless Tubing Nuts, Chemglass
Catalog # 80060-480
Supplier:  Chemglass
Supplier Number:  CG-1164-N-02
Small Business Enterprise
Specifications
  • Description:
    Tefzel® Flangeless Nut, 1/8"
  • Cat. no.:
    80060-480
  • Supplier No.:
    CG-1164-N-02
Specifications
About this item
Flangeless nut eliminates the need to flange tubing.
  • Use with a flangeless ferrule for a leak-free connection of FEP tubing
  • Nut is molded out of Tefzel®
  • Nut has a 1/4"–28 thread
Simply insert tubing through the nut, attach the ferrule (flat end towards the PTFE fitting) to the end of the tubing, and finger tighten the nut into place. Nut has a 1/4"–28 thread, and is molded out of natural Tefzel®, a soft fluoropolymer resistant to chemical attack.