About this item
Compact laboratory water systems for 10 liters per day.
- Reliable: Delivers consistently high water quality for reliable and reproducible results
- Intuitive: Touch-activated color display with direct access to all important dispensing options
- Innovative: With unique bagtank technology, depending on the type of system; saves time-intensive tank cleaning
- Compact: Space-saving, with a width of only 28 cm
- Water purification method adsorption by spherical activated carbon, catalyst, reverse osmosis, ion exchange, optional UV irradiation, and by point-of-use Particle-removing filtration/sterile filtration
Reliable to ensure that you always obtain reliable and reproducible results, the system ensures consistently high water quality. For your analytical requirements and especially critical applications, you additionally have the option of obtaining your system with an integrated UV lamp (185/254 nm) to reduce TOC to <5 ppb*. The most advanced bagtank technology will save you from performing time-intensive cleaning and rinsing procedures. As this eliminates the need for using chemicals that can be hazardous to your health, you will help protect the environment and increase your own safety. Easily operate the arium® mini using the touch-activated color display – even when you are wearing laboratory gloves. Easy-tounderstand icons will guide you through the menu for intuitive, error-free operation. Simplify your sample preparation and benefit from direct access to all important dispensing functions: manual, volume-controlled or predefined volumes. With a width of only 28 cm, arium® mini will readily fit into any laboratory environment. This handy device will give you the flexibility you need in integrating it into nearly any location.
Specifications
- Catalog No:
- 76290-370
- 76290-372
- Model:
- Arium® mini
- Arium® mini UV
- TOC content:
- 10 ppb
- 5 ppb
- Resistivity at +25 °C:
- 18.2 MΩ × cm
- 18.2 MΩ × cm
- Electrical:
- 100 – 240 VAC; 50 and 60 Hz, 2 A (max.)
- 100 – 240 VAC; 50 and 60 Hz, 2 A (max.)
- W×D×H:
- 280×509.4×530.7 mm (11×20×20.9")
- 280×509.4×530.7 mm (11×20×20.9")
- Weight:
- Approx. 13 kg (28.6 lbs.)
- Approx. 13 kg (28.6 lbs.)