Economical Class 7 Hooked Weights
About this item
For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.
- Weights above 5 g are constructed of 303 Stainless Steel
- Weights 5 g and below are constructed of 3003-H14 Aluminum
- Manufactured in the USA
All weights are hooked and feature two-piece construction. Weight sets are supplied in a weight block with compartments for each weight. Individual weights are not supplied in a case.
8 Options Available Below
Product Details & Documents
For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.
- Weights above 5 g are constructed of 303 Stainless Steel
- Weights 5 g and below are constructed of 3003-H14 Aluminum
- Manufactured in the USA
All weights are hooked and feature two-piece construction. Weight sets are supplied in a weight block with compartments for each weight. Individual weights are not supplied in a case.
Product Family Options
Weight Sets
Product Information
- No. of weightsAvailabilityPrice
- 470204-266Hooked Weights, 10 g to 1000 gSupplier Number: 30390775
- 470013-386Hooked Weights, 0.1 g to 10 gSupplier Number: 30390785
Individual Weights
Product Information
- No. of weightsAvailabilityPrice
- 470013-388Hooked Weight, 10 gSupplier Number: 30390792
- 470013-438Hooked Weight, 20 gSupplier Number: 30390791
- 470013-390Hooked Weight, 50 gSupplier Number: 30390790
- 470017-682Hooked Weight, 200 gSupplier Number: 30390788
- 470017-684Hooked Weight, 500 gSupplier Number: 30390787
- 470017-686Hooked Weight, 1000 gSupplier Number: 30390786