Chromacol™ GOLD-Grade Inert Vials and Inserts, Thermo Scientific
GOLD-Grade products offer Ultra low absorption and excellent recovery for trace analysis.
- lnert glass vials
- Special 33 expansion glass
- Ultra-low pH shift
- Stable analysis and higher reproducibility
- Ultra-low adsorption surface for critical polar compounds
- Highest reproducibility
With uniform wall thickness; white enamel, double-graduated metric scale; and a marking spot, the vials can be heated and used with acids, salt solutions, and organic solvents.