Alpha1-2,3,6 Mannosidase
Catalog # CA103258-582
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Size:80 units
- Enzyme name:alpha1-2,3,6 Mannosidase
- Concentration:2000 units/ml
- Cat. no.:CA103258-582
Specifications
About this item
Recombinant enzyme with no detectable endoglycosidase or other exoglycosidase contaminating activity.
- Glycerol-free for optimal performance in HPLC and mass spectrometry analysis
- ≥95% purity, as determined by SDS-PAGE and intact ESI-MS
- Optimal activity and stability for up to 12 months
1-2,3,6 Mannosidase, cloned from Jack Bean, and also known as JBM, is a broad specificity exoglycosidase that catalyzes the hydrolysis of terminal 1-2, 1-3 and 1-6 linked mannose residues from oligosaccharides. 1-2,3,6 Mannosidase has a slight preference for 1-2 mannose residues over 1-3 and 1-6 mannose residues.