Bioruptor® Plus TPX Microtubes, Diagenode
MSPP-C30010010
- Description:Bioruptor® Plus TPX microtubes
- Size:1.5 ml
- Cat. no.:MSPP-C30010010
- Packaging:300/pack
- Supplier No.:C30010010-50
TPX® Polymethylpentene (PMP) tubes are designed to improve sonication and shearing efficiency.
- Recommended sample volume: 100 to 300 μl
TPX® is a hard plastic which improves the transmission of the ultrasound to the sample. These tubes have been validated to be used with Bioruptor® Standard and Bioruptor® Plus ultrasonicators.