Order Entry
Canada
ContactUsLinkComponent
BeadBeater Laboratory Homogenizer, Biospec Products
Catalogue # MSPP-1107900
Fournisseur:  BIOSPEC PRODUCTS MS
BeadBeater Laboratory Homogenizer, Biospec Products
Catalogue # MSPP-1107900
Fournisseur:  BIOSPEC PRODUCTS MS
Supplier Number:  1107900

Spécifications

  • Description:
    BeadBeater
  • Dimensions:
    6Ø×12"H
  • Cat. no.:
    MSPP-1107900
  • Electrical:
    115/220 - 240 V, 50/60 Hz
  • Speed Range:
    15800 rpm
  • Sample Volume:
    200 ml
  • Volume Range:
    350 ml

Spécifications

À propos de cet article

The BeadBeater is the ultimate laboratory-scale homogenizer for disrupting aqueous suspensions of yeast, fungi or bacteria. Complete cell lysis of 10 to 80 g (wet weight) of cells is achieved in about three minutes of operation.

  • Ideal for isolating proteins, enzymes and intact intracellular organelles
  • Sample cooling during homogenisation is simple and efficient
  • Powerful, high speed motor
  • Only durable, non-toxic components will be contact the product
  • Designed for batches up to 80 g (wet weight) in 200 ml of medium
  • Highly efficient disruption in shortest possible times
  • Homogeniser clean-up and mantinance is uncomplicated and easy
Delivery: The BeadBeater is supplied with a 350 ml polycarbonate chamber, rotor assembly, motor base, ice-water cooling jacket, and one pound of glass beads of your choice.
Caution: Glass beads and zirconia-silica beads having diameters of 0.1 mm, 0.5 mm and 1 mm are the prefered grinding media. Larger and heavier beads such as steel beads cannot be adequately agitated.