Ammonia solution 10.0 N volumetric solution
Polynatural, 500 ml
Supplier: Ricca Chemical
: R0647000-500A
Ammonia solution 10.0 N volumetric solution
Catalog # RCR0647000500A
Supplier: Ricca Chemical
CAS Number: 1336-21-6
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- UN:2672
- ADR:8,III
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Assay (vs. sulfuric acid/methyl red):9.95 - 10.05 N at 20 °C
Specifications
- Pack type:Polynatural
- Size:500 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



