Orders LinkContactUsLinkComponent
Dispensing Tips, EFD INC.
Dispensing Tips, EFD INC.
Certificates
Dispensing Tips, EFD INC.
2 Options Available Below
Product Details & Documents
Smooth flow for application of medium- to high-viscosity fluids especially thick or particle-filled materials like epoxies, RTVs and braze pastes.
  • Free of flash, burrs and contaminants
  • Package labels include lot numbers for process control and traceability
  • Consistent from style to style and lot to lot
  • 360° SafetyLok™ thread ensures safe, positive attachment to syringe barrel
  • Engineered hub flats for easy twist on, twist off
Molded of polyethylene with UV-light block additive.
 : Packaged 50 tips per box.
Product Family Options
Product Information
  • Gauge
    Point style
    Color
    Availability
    Price
  • 16
    Opaque Rigid
    Grey
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 50
    Box of 50 - $130.72
    $130.72
    Log in for availability
    Each
    Each - $130.72
    $130.72
    Box of 50
       $130.72
    Each
       $130.72
    Supplier Number: 7018097
  • 25
    Standard
    Red
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $260.39
    $260.39
    Log in for availability
    Pack of 50
    Pack of 50 - $261.49
    $261.49
    Each
       $260.39
    Pack of 50
       $261.49
    Supplier Number: 7018391