ContactUsLinkComponent
Tech-Med® Perforated Insert Trays, DUKAL Corporation®
Tech-Med® Perforated Insert Trays, DUKAL Corporation®
Catalog # 76235-478
Supplier:  Dukal
undefined
Tech-Med® Perforated Insert Trays, DUKAL Corporation®
Catalog # 76235-478
Supplier:  Dukal
Supplier Number:  4276P
Specifications
  • Description:
    Perforated Insert Tray for use with 4276
  • Dimensions:
    10.25×6.25×4"
  • Material:
    Stainless steel
  • Cat. no.:
    76235-478
Specifications
About this item
Trays are crafted without seams to prevent bacteria growth and allow for easy cleaning.
  • Made of the highest grade stainless steel (18/8)
  • Autoclavable