
Hydrochloric acid 10 g/L HCl
Catalog # RCR3563000-1A
Supplier: Ricca Chemical
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. no.:RCR3563000-1A
Specifications
Specification Test Results
- Appearance:Colorless liquid
- Assay (vs. Sodium Hydroxide/Phenolphthalein):9.9-10.1 g/L
Specification Test Results
About this item
:
:36.46
:GHS08
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324