ContactUsLinkComponent
Mini-Run Electrophoresis System, Bulldog Bio
Mini-Run Electrophoresis System, Bulldog Bio
Catalog # 102408-078
Supplier:  Bulldog Bio
undefined
Mini-Run Electrophoresis System, Bulldog Bio
Catalog # 102408-078
Supplier:  Bulldog Bio
Supplier Number:  BMR100
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Mini-Run Electrophoresis System
  • Cat. no.:
    102408-078
  • Supplier No.:
    BMR100
Specifications
About this item
Down size and pay less.
The Mini-Run horizontal gel apparatus can be used for almost any agarose gel electrophoresis application. The chamber has a built-in DC power supply so you can just plug it in and be ready to run your gel.
The cover and gel chamber are both made from an advanced etch-resistant plastic and the electrodes are platinum coated for clean operation and durability.