Order Entry
Canada
ContactUsLinkComponent
Interleukin-6 (Mouse) ELISA Kit, Cayman Chemical
Interleukin-6 (Mouse) ELISA Kit, Cayman Chemical
Catalog # 75816-960
CAS Number:  
undefined
Interleukin-6 (Mouse) ELISA Kit, Cayman Chemical
Catalog # 75816-960
Supplier Number:  583371-96
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Interleukin-6 (mouse) ELISA Kit
  • Cat. no.:
    75816-960
  • Contents:
    Kit contains: ELISA Strip Plate, Biotin Conjugate, ELISA Standard, Streptavidin-HRP, Immunoassay Buffer B Concentrate, Polysorbate 20, 96-Well Cover Sheet, Substrate Solution, Wash Buffer Concentrate, and Stop Solution
  • Supplier No.:
    583371-96
  • No. of Tests:
    96 wells

Specifications

About this item

Enzyme immunometric assay for the measurement of mouse IL-6.

  • Measure IL-6 from mouse plasma, serum, and other sample matrices
  • Rapid assay
  • Get results in under 3 hours

Interleukin-6 (IL-6) is produced by monocytes, macrophages, and T cells and is invovled in many defense mechanisms including the immune response, hematopoiesis, bone metabolism, and acute phase reactions.

Cayman’s IL-6 (mouse) ELISA Kit is an immunometric (sandwich) ELISA that permits IL-6 measurements within the range of 23.4 to 1500 pg/ml, with an LLOQ of approximately 23.4 pg/ml.