bisBenzimide H 33258 trihydrochloride (Hoechst 33258) 20 mM in water
Catalog # 103011-144
Supplier: Anaspec Inc
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:5 ml
- Cat. no.:103011-144
- Supplier No.:AS-83219
Specifications
About this item
:
:533.88 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:23491-45-4
Formula:C₂₅H₂₄N₆O·3HCl
MDL Number:MFCD00012679