Orders LinkContactUsLinkComponent
Sharp-Point Surgical Scissors
Sharp-Point Surgical Scissors, 140 mm (5.5"), Stainless steel, Sharp/Sharp
  470105-650
 :  22272
Sharp-Point Surgical Scissors
  470105-650
Sharp-Point Surgical Scissors
Sharp-Point Surgical Scissors
Sharp-Point Surgical Scissors, 140 mm (5.5"), Stainless steel, Sharp/Sharp
$10.65
Each Case of 25
Start Your Cuts Easily.
  • Dissect without a scalpel
  • Handles rounded for ambidextrous use
Sharp-pointed, straight surgical scissors allow the user to insert blades into tissue to start the cut. Larger scissors provide greater leverage for tough cutting; shorter scissors for finer work. Stainless steel.
  • Description:
    Sharp-Point Surgical Scissors
  • Length:
    140 mm (5.5")
  • Material:
    Stainless steel
  • Blade type:
    Sharp/Sharp