Order Entry
Canada
ContactUsLinkComponent
 
Total RNA Isolation Mini Kit, Agilent Technologies
  CA97066-614
 :  
undefined
Total RNA Isolation Mini Kit, Agilent Technologies
  CA97066-614
 :  5185-6000
 :  

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    Total RNA Isolation Mini Kit
  • Description:
    Total RNA Isolation Mini Kit
  • Cat. no.:
    CA97066-614

 

 

RNA isolation kits provide a straightforward, spin-column method to deliver RNA for use in gene expression and other downstream analysis.