About this item
The ZAP Express and the Lambda ZAP-CMV vectors combine the high efficiency of lambda vector systems with the versatility of plasmid systems.
The ZAP Express vectors enable both prokaryotic and eukaryotic expression of gene of interest.
In vivo excision of the ZAP Express vector yields the PBK-CMV vector.
The Lambda ZAP-CMV vector enables high-level eukaryotic expression of gene of interest. In vivo excision of the lambda ZAP-CMV vector yields the pCMV-Script-EX vector.
The CMV immediate early promoter drives gene expression in mammalian cells.
The lacZ promoter drives prokaryotic expression, while the CMV immediate early promoter drives gene expression in mammalian cells.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Related Information
Manual ExAssist Interference-Resistant Helper Phage with SOLR Strain Manual ExAssist Interference-Resistant Helper Phage with XLOLR Strain Manual Lambda ZAP-CMV XR Predigested Vector Kit Manual Rapid Excision Kit Manual ZAP Express Predigested Vector Kit and ZAP Express Predigested Gigapack Cloning Kits