Order Entry
Canada
ContactUsLinkComponent
Human Recombinant alpha-Synuclein (from E. coli)
Human Recombinant alpha-Synuclein (from E. coli)
Catalog # 102890-616
Supplier:  R&D Systems
CAS Number:  
Human Recombinant alpha-Synuclein (from E. coli)
Catalog # 102890-616
Supplier:  R&D Systems
Supplier Number:  SP-480-500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    500 μg
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 6 months, -70 °C under sterile conditions after opening.
  • Gene ID:
    6622
  • Protein synonyms:
    PARK1|alpha (non A4 component of amyloid precursor)|SNCA|PARK4|Lewy body) 4|Non-A beta component of AD amyloid|Synuclein-alpha|Non-A4 component of amyloid precursor|MGC110988|NACP|non-A4 component of amyloid|synuclein|non A-beta component of AD amyloid
  • Accession no.:
    P37840
  • Protein/peptide name:
    alpha-Synuclein
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain.
  • Cat. no.:
    102890-616
  • Supplier No.:
    SP-480-500

Specifications

About this item

The Recombinant Human His6-Synuclein-alpha Protein from R&D Systems, powered by R&D Systems, powered by Boston Biochem is derived from E. coli. The Recombinant Human His6-Synuclein-alpha Protein has been validated for the following applications: Bioactivity.