Order Entry
Canada
ContactUsLinkComponent
Human Recombinant UAF1/WDR48 (from Baculovirus (Sf21 Insect Cells))
Human Recombinant UAF1/WDR48 (from Baculovirus (Sf21 Insect Cells))
Catalog # 102880-874
Supplier:  R&D Systems
CAS Number:  
Human Recombinant UAF1/WDR48 (from Baculovirus (Sf21 Insect Cells))
Catalog # 102880-874
Supplier:  R&D Systems
Supplier Number:  E-566-050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus (Sf21 Insect cells)
  • Species:
    Human
  • Size:
    50 μg
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening.
  • Gene ID:
    57599
  • Protein synonyms:
    WD repeat endosomal protein|P80|WD repeat domain 48|USP1-associated factor 1|WDR48|KIAA1449DKFZp686G1794|UAF1|WD repeat-containing protein 48|USP1 associated factor 1
  • Accession no.:
    Q8TAF3
  • Protein/peptide name:
    UAF1/WDR48
  • Purity:
    >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain
  • Cat. no.:
    102880-874
  • Supplier No.:
    E-566-050

Specifications

About this item

The Recombinant Human His6-UAF1 Protein from R&D Systems, powered by R&D Systems, powered by Boston Biochem is derived from Sf 21 (baculovirus). The Recombinant Human His6-UAF1 Protein has been validated for the following applications: Bioactivity.