Order Entry
Canada
ContactUsLinkComponent
Human Recombinant UBE2F/NCE2 (from E. coli)
Human Recombinant UBE2F/NCE2 (from E. coli)
Catalog # 102880-598
Supplier:  R&D Systems
CAS Number:  
Human Recombinant UBE2F/NCE2 (from E. coli)
Catalog # 102880-598
Supplier:  R&D Systems
Supplier Number:  E2-679-100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 μg
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening.
  • Gene ID:
    140739
  • Protein synonyms:
    EC 6.3.2.-|ubiquitin-conjugating enzyme E2F (putative)|NEDD8-conjugating enzyme 2|NEDD8 protein ligase UBE2F|NCE2Ubiquitin-conjugating enzyme E2 F|NEDD8 carrier protein UBE2F|NCE2|NEDD8 conjugating enzyme|MGC18120|NEDD8-conjugating enzyme UBE2F|EC 6.3.2
  • Accession no.:
    NP_542409
  • Protein/peptide name:
    UBE2F/NCE2
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain
  • Cat. no.:
    102880-598
  • Supplier No.:
    E2-679-100

Specifications

About this item

The Recombinant Human His6-UBE2F Protein from R&D Systems, powered by R&D Systems, powered by Boston Biochem is derived from E. coli. The Recombinant Human His6-UBE2F Protein has been validated for the following applications: Enzyme Activity.