Human Recombinant UbcH5c/UBE2D3 (from E. coli)
102880-444
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening.
- Gene ID:7323
- Protein synonyms:ubiquitin-conjugating enzyme E2D 3 (UBC4/5 homolog|Ubiquitin-conjugating enzyme E2-17 kDa 3|UBC5C|MGC5416|E2(17)KB3|Ubiquitin-protein ligase D3|MGC43926|Ubiquitin-conjugating enzyme E2(17)KB 3|ubiquitin-conjugating enzyme E2 D3|UBCH5C|UBC4/5|ubiquitin-conjugating enzyme E2D 3 (homologous to yeast UBC4/5)|Ubiquitin carrier protein D3|yeast)|E2(17)KB 3|EC 6.3.2.19|UBE2D3
- Accession no.:P61077
- Protein/peptide name:UbcH5c/UBE2D3
- Purity:>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain
- Cat. no.:102880-444
- Supplier No.:E2-627-100
The Recombinant Human UbcH5c/UBE2D3 Protein from R&D Systems, powered by R&D Systems, powered by Boston Biochem is derived from E. coli. The Recombinant Human UbcH5c/UBE2D3 Protein has been validated for the following applications: Enzyme Activity.