Mouse Recombinant SEMA3D (from CHO cells)
Catalog # MSPP-9386S3025
Supplier: R&D Systems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Size:25 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin content:<0.10 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:223117
- Protein synonyms:coll-2|short basic domain|semaphorin-3D|Sema-Z2|immunoglobulin domain (Ig)|MGC39708|secreted|SEMA3D|sema domain|(semaphorin) 3D|Collapsin 2
- Accession no.:Q8BH34
- Protein/peptide name:SEMA3D
- Purity:>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
- Molecular weight:97 - 113 kDa, reducing conditions
- Cat. no.:MSPP-9386S3025
Specifications
About this item
The Recombinant Mouse Semaphorin 3D Fc Chimera Protein from R&D Systems is derived from CHO. The Recombinant Mouse Semaphorin 3D Fc Chimera Protein has been validated for the following applications: Bioactivity.