Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin content:<0.10 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:23308
- Protein synonyms:ICOS-L|B7-H2B7-related protein 1|B7RP1B7-like protein Gl50|inducible T-cell co-stimulator ligand|B7RP-1|ICOSLG|CD275|B7RP-1LICOS|GL50transmembrane protein B7-H2 ICOS ligand|CD275 antigen|KIAA0653ICOS ligand|ICOSL|GL50|B7H2B7 homolog 2|B7H2
- Accession no.:O75144
- Protein/peptide name:B7-H2/ICOSLG
- Purity:>95%, by SDS-PAGE with silver staining.
- Molecular weight:59-64 kDa, reducing conditions
- Cat. no.:102869-372
- Supplier No.:8206-B7-100
The Recombinant Human B7-H2 His-tag Protein from R&D Systems is derived from HEK293. The Recombinant Human B7-H2 His-tag Protein has been validated for the following applications: Bioactivity.