Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant CD177 (from NS0 cells)
Mouse Recombinant CD177 (from NS0 cells)
Catalog # 102869-364
Supplier:  R&D Systems
CAS Number:  
Mouse Recombinant CD177 (from NS0 cells)
Catalog # 102869-364
Supplier:  R&D Systems
Supplier Number:  8186-CD-050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    NS0 cells
  • Species:
    Mouse
  • Size:
    50 μg
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    57126
  • Protein synonyms:
    CD177|HNA-2a|NB1 GP|HNA2Acell surface receptor|Human neutrophil alloantigen 2a|polycythemia rubra vera 1|CD177 molecule|PRV1|CD177 antigenNB1 GP|PRV1NB1 glycoprotein|Polycythemia rubra vera protein 1|NB1PRV-1
  • Accession no.:
    Q8R2S8
  • Protein/peptide name:
    CD177
  • Purity:
    >95%, by SDS-PAGE with silver staining
  • Molecular weight:
    100-115 kDa, reducing conditions
  • Cat. no.:
    102869-364
  • Supplier No.:
    8186-CD-050

Specifications

About this item

The Recombinant Mouse CD177 Protein from R&D Systems is derived from NS0. The Recombinant Mouse CD177 Protein has been validated for the following applications: Bioactivity.