Order Entry
Canada
ContactUsLinkComponent
 
Rat Recombinant CD27/TNFRSF7 (from NS0 cells)
Catalog # 102877-284
Supplier:  R&D Systems
CAS Number:  
Rat Recombinant CD27/TNFRSF7 (from NS0 cells)
Catalog # 102877-284
Supplier:  R&D Systems
Supplier Number:  7888-CD-050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    NS0 cells
  • Species:
    Rat
  • Size:
    50 μg
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    939
  • Protein synonyms:
    Tp55|CD27 antigen|TNFRSF7T-cell activation antigen CD27|member 7|CD27 molecule|S152CD27L receptor|tumor necrosis factor receptor superfamily|Tumor necrosis factor receptor superfamily member 7|MGC20393|T14|TNFRSF7|CD27|T cell activation antigen S152|T cell activation antigen CD27
  • Accession no.:
    NP_001019506
  • Protein/peptide name:
    CD27/TNFRSF7
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Molecular weight:
    60-66 kDa, reducing conditions
  • Cat. no.:
    102877-284
  • Supplier No.:
    7888-CD-050

Specifications

About this item

The Recombinant Rat CD27/TNFRSF7 Fc Chimera Protein from R&D Systems is derived from NS0. The Recombinant Rat CD27/TNFRSF7 Fc Chimera Protein has been validated for the following applications: Bioactivity.