Mouse Recombinant Ephrin-A3 (from NS0 Cells)
Catalog # 102868-858
Supplier: R&D Systems
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:NS0 cells
- Species:Mouse
- Size:50 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin content:<0.10 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:1944
- Protein synonyms:Ehk1-L|EPLG3EHK1 ligand|EphrinA3|EFL2|LERK3|EPH-related receptor tyrosine kinase ligand 3|ephrin-A3|LERK3LERK-3|EFL-2|ligand of eph-related kinase 3|EFNA3
- Accession no.:NP_034238
- Protein/peptide name:Ephrin-A3
- Purity:>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular weight:60-66 kDa, reducing conditions
- Cat. no.:102868-858
- Supplier No.:7395-EA-050
Specifications
About this item
The Recombinant Mouse Ephrin-A3 Fc Chimera Protein from R&D Systems is derived from NS0. The Recombinant Mouse Ephrin-A3 Fc Chimera Protein has been validated for the following applications: Bioactivity.