Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:25 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin content:<0.10 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:4041
- Protein synonyms:low density lipoprotein receptor-related protein 7|LR3VBCH2|LRP7|EVR4|OPS|OPPG|BMND1|HBM|BMND1OPTA1|osteoporosis pseudoglioma syndrome|low-density lipoprotein receptor-related protein 5|low density lipoprotein receptor-related protein 5|VBCH2|LRP5|EVR1|OPTA1|LRP7exudative vitreoretinopathy 1|LRP-5
- Accession no.:NP_032539
- Protein/peptide name:LRP-5
- Purity:>85%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular weight:160 kDa, reducing conditions
- Cat. no.:102868-824
- Supplier No.:7344-LR-025
The Recombinant Mouse LRP-5 Protein from R&D Systems is derived from HEK293. The Recombinant Mouse LRP-5 Protein has been validated for the following applications: Bioactivity.