Order Entry
Canada
ContactUsLinkComponent
Human Recombinant ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 (from CHO Cells)
Human Recombinant ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 (from CHO Cells)
Catalog # 102868-804
Supplier:  R&D Systems
CAS Number:  
Human Recombinant ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 (from CHO Cells)
Catalog # 102868-804
Supplier:  R&D Systems
Supplier Number:  7275-GT-020
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    20 μg
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
  • Endotoxin content:
    <1.0 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    6483
  • Protein synonyms:
    Gal-NAc6S|EC 2.4.99.4|ST3 beta-galactoside alpha-2,3-sialyltransferase 2|sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase)|Alpha 2,3-ST 2|Sialyltransferase 4B|ST3GalA.2ST3Gal II|CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2|SIAT4-B|Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase|Beta-galactoside alpha-2,3-sialyltransferase 2|SIAT4B|EC 2.4.99.7|ST3GalA.2|beta-galactoside alpha-2,3-sialytransferase|ST3GALII
  • Accession no.:
    NP_008858
  • Protein/peptide name:
    ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain at 5 ╡g per lane
  • Molecular weight:
    30-40 kDa, reducing conditions
  • Cat. no.:
    102868-804
  • Supplier No.:
    7275-GT-020

Specifications

About this item

The Recombinant Human ST3GAL2 Protein from R&D Systems is derived from CHO. The Recombinant Human ST3GAL2 Protein has been validated for the following applications: Enzyme Activity.