Mouse Recombinant sFRP-5 (from CHO cells)
Catalog # 102868-784
Supplier: R&D Systems
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Size:50 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin content:<0.01 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:6425
- Protein synonyms:SARP3|secreted apoptosis related protein 3|secreted frizzled-related protein 5|frizzled-related protein 1b|secreted apoptosis-related protein 3|sFRP5|FRP-1b|sFRP-5|SARP-3|FRP1B
- Accession no.:NP_061250
- Protein/peptide name:sFRP-5
- Purity:>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular weight:39-42 kDa, reducing conditions
- Formulation:Lyophilized from a 0.2 μm filtered solution in PBS and 400 mM NaCl, pH 6.8 with 5% Trehalose.
- Cat. no.:102868-784
- Supplier No.:7195-SF-050
Specifications
About this item
The Recombinant Mouse sFRP-5 Protein from R&D Systems is derived from CHO. The Recombinant Mouse sFRP-5 Protein has been validated for the following applications: Bioactivity.
Measured by its ability to inhibit Wnt induced TCF reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 0.600-7.20 μg/ml.