Human Recombinant VISTA/B7-H5/PD-1H (from NS0 Cells)
Catalog # MSPP-7126B701M
Supplier: R&D Systems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:NS0 cells
- Species:Human
- Size:1 mg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin content:<0.01 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:64115
- Protein synonyms:B7-H5|C10orf54|PP2135|SISP1|VSIR|platelet receptor Gi24|Gi24|chromosome 10 open reading frame 54|stress induced secreted protein 1|B7H5|PD-1H|PD1H|Dies1|4632428N05Rik
- Accession no.:AAH20568
- Protein/peptide name:VISTA/B7-H5/PD-1H
- Purity:>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
- Molecular weight:64-75 kDa, reducing conditions
- Cat. no.:MSPP-7126B701M
Specifications
About this item
The Recombinant Human VISTA/B7-H5/PD-1H Fc Chimera Protein from R&D Systems is derived from NS0. The Recombinant Human VISTA/B7-H5/PD-1H Fc Chimera Protein has been validated for the following applications: Bioactivity.