Human Recombinant Adenosine Deaminase/ADA (from Baculovirus (Sf21 Insect cells))
102867-670
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Baculovirus (Sf21 Insect cells)
- Species:Human
- Size:10 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
- Endotoxin content:<1.0 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:100
- Protein synonyms:ADA|Adenosine aminohydrolase|ADA1|EC 3.5.4.4|adenosine deaminase|adenine deaminase
- Accession no.:P00813
- Protein/peptide name:Adenosine Deaminase
- Purity:>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
- Molecular weight:40-42 kDa, reducing conditions
- Cat. no.:102867-670
- Supplier No.:7048-AD-010
The Recombinant Human Adenosine Deaminase/ADA Protein from R&D Systems is derived from Sf 21 (baculovirus). The Recombinant Human Adenosine Deaminase/ADA Protein has been validated for the following applications: Enzyme Activity.