Rat Recombinant CCL4/MIP-1 beta (from E. coli)
102868-740
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Size:25 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin content:<0.10 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:6351
- Protein synonyms:MIP-1-beta(1-69)|small inducible cytokine A4 (homologous to mouse Mip-1b)|SCYA4|Macrophage inflammatory protein 1-beta|HC21|ACT-2|G-26|PAT 744|Small-inducible cytokine A4|AT744.1|LAG-1|MIP-1-beta|SCYA2|MIP1 beta|SIS-gamma|G-26 T-lymphocyte-secreted protein|C-C motif chemokine 4|ACT2|Lymphocyte activation gene 1 protein|Protein H400|MIP-1 beta|MIP1B|MIP1B1|T-cell activation protein 2|LAG1|chemokine (C-C motif) ligand 4|Exodus-3|secreted protein G-26
- Accession no.:P50230
- Protein/peptide name:CCL4/MIP-1 beta
- Purity:>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular weight:9 kDa, reducing conditions
- Cat. no.:102868-740
- Supplier No.:6935-MB-025
The Recombinant Rat CCL4/MIP-1 beta Protein from R&D Systems is derived from E. coli. The Recombinant Rat CCL4/MIP-1 beta Protein has been validated for the following applications: Bioactivity.