Order Entry
Canada
ContactUsLinkComponent
Influenza A H3N2 Recombinant Neuraminidase (from Baculovirus (Sf21 Insect cells))
Influenza A H3N2 Recombinant Neuraminidase (from Baculovirus (Sf21 Insect cells))
Catalog # 102868-722
Supplier:  R&D Systems
CAS Number:  
Influenza A H3N2 Recombinant Neuraminidase (from Baculovirus (Sf21 Insect cells))
Catalog # 102868-722
Supplier:  R&D Systems
Supplier Number:  6875-NM-005
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Enzyme type:
    Recombinant
  • Source:
    Baculovirus (Sf21 Insect cells)
  • Species:
    Influenza A H3N2
  • Size:
    5 μg
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70 C as supplied. 3 months, -70 C under sterile conditions after opening.
  • Endotoxin content:
    <1.0 EU per 1 ╡g of the protein by the LAL method.
  • Enzyme name:
    Neuraminidase
  • Purity:
    >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain at 5 ╡g per lane
  • Molecular weight:
    57 - 65 kDa, reducing conditions
  • Cat. no.:
    102868-722
  • Supplier No.:
    6875-NM-005

Specifications

About this item

The Recombinant Influenza A Virus H3N2 Neuraminidase Protein from R&D Systems is derived from Sf 21 (baculovirus). The Recombinant Influenza A Virus H3N2 Neuraminidase Protein has been validated for the following applications: Enzyme Activity.

  • Accession number: Q91MA2