Order Entry
Canada
ContactUsLinkComponent
Human Recombinant sFRP-2 (from NS0 Cells)
Human Recombinant sFRP-2 (from NS0 Cells)
Catalog # 102868-716
Supplier:  R&D Systems
CAS Number:  
Human Recombinant sFRP-2 (from NS0 Cells)
Catalog # 102868-716
Supplier:  R&D Systems
Supplier Number:  6838-FR-025/CF
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    NS0 cells
  • Species:
    Human
  • Size:
    25 μg
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    6423
  • Protein synonyms:
    secreted frizzled-related protein 2|FRP2|SARP-1|sFRP2|SARP1sFRP-2|SDF-5|Secreted apoptosis-related protein 1|FRP-2secreted apoptosis related protein 1
  • Accession no.:
    AAH08666
  • Protein/peptide name:
    sFRP-2
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Molecular weight:
    34-36 kDa, reducing conditions
  • Cat. no.:
    102868-716
  • Supplier No.:
    6838-FR-025/CF

Specifications

About this item

The Recombinant Human sFRP-2 Protein from R&D Systems is derived from NS0. The Recombinant Human sFRP-2 Protein has been validated for the following applications: Bioactivity.