Order Entry
Canada
ContactUsLinkComponent
Human Recombinant PlGF-2 (from CHO Cells)
Human Recombinant PlGF-2 (from CHO Cells)
Catalog # 102868-712
Supplier:  R&D Systems
CAS Number:  
Human Recombinant PlGF-2 (from CHO Cells)
Catalog # 102868-712
Supplier:  R&D Systems
Supplier Number:  6837-PL-025
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    25 μg
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin content:
    <0.01 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    5281
  • Accession no.:
    NP_002623
  • Protein/peptide name:
    PlGF-2
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Molecular weight:
    28-33 kDa, reducing conditions
  • Cat. no.:
    102868-712
  • Supplier No.:
    6837-PL-025

Specifications

About this item

The Recombinant Human PlGF-2 Protein from R&D Systems is derived from CHO. The Recombinant Human PlGF-2 Protein has been validated for the following applications: Bioactivity.