Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Baculovirus (Sf21 Insect cells)
- Species:Human
- Size:10 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
- Endotoxin content:<1.0 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:5321
- Protein synonyms:phosphatidylcholine 2-acylhydrolase|cPLA2|calcium-dependent phospholipid-binding protein|Phospholipase A2 group IVA|phospholipase A2|PLA2G4cytosolic phospholipase A2|calcium-dependent)|MGC126350|group IVA (cytosolic|cPLA2-alpha|lysophospholipase
- Accession no.:NP_077734
- Protein/peptide name:PLA2G4A
- Purity:>80%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
- Molecular weight:85-95 kDa, reducing conditions
- Cat. no.:102868-610
- Supplier No.:6659-PL-010
The Recombinant Human PLA2G4A Protein from R&D Systems is derived from Sf 21 (baculovirus). The Recombinant Human PLA2G4A Protein has been validated for the following applications: Enzyme Activity.