Order Entry
Canada
ContactUsLinkComponent
Rat Recombinant CCL3/MIP-1 alpha (from E. coli)
Rat Recombinant CCL3/MIP-1 alpha (from E. coli)
  102877-154
 :  
Rat Recombinant CCL3/MIP-1 alpha (from E. coli)
  102877-154
 :  6625-MA-025/CF
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Size:
    25 μg
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    6348
  • Protein synonyms:
    AI323804|G0S19-1|MIP1-(a)|Scya3|MIP1 alpha|LD78alpha|chemokine (C-C motif) ligand 3|MIP-1alpha|MIP1-alpha|C-C motif chemokine 3|LD78a|MIP-1 alpha|Mip1a
  • Accession no.:
    P50229
  • Protein/peptide name:
    CCL3/MIP-1 alpha
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Molecular weight:
    8.5 kDa, reducing conditions
  • Cat. no.:
    102877-154
  • Supplier No.:
    6625-MA-025/CF

 

 

The Recombinant Rat CCL3/MIP-1 alpha Protein from R&D Systems is derived from E. coli. The Recombinant Rat CCL3/MIP-1 alpha Protein has been validated for the following applications: Bioactivity.