Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Biliverdin Reductase B/BLVRB (from E. coli)
Human Recombinant Biliverdin Reductase B/BLVRB (from E. coli)
Catalog # 102877-124
Supplier:  R&D Systems
CAS Number:  
Human Recombinant Biliverdin Reductase B/BLVRB (from E. coli)
Catalog # 102877-124
Supplier:  R&D Systems
Supplier Number:  6568-BR-020
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    20 μg
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
  • Endotoxin content:
    <1.0 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    645
  • Protein synonyms:
    FR|NADPH-flavin reductase|GHBP|Green heme-binding protein|short chain dehydrogenase/reductase family 43U|FLRBVRB|EC 1.3.1.24|biliverdin reductase B (flavin reductase (NADPH))|BVR-B|BLVRB|Biliverdin reductase B|flavin reductase (NADPH)|Biliverdin-IX beta-reductase|member 1|SDR43U1|NADPH-dependent diaphorase|EC 1.5.1.30|FLR|MGC117413|flavin reductase|BVRB
  • Accession no.:
    P30043
  • Protein/peptide name:
    Biliverdin Reductase B/BLVRB
  • Purity:
    >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
  • Molecular weight:
    23-25 kDa, reducing conditions
  • Cat. no.:
    102877-124
  • Supplier No.:
    6568-BR-020

Specifications

About this item

The Recombinant Human Biliverdin Reductase B/BLVRB Protein from R&D Systems is derived from E. coli. The Recombinant Human Biliverdin Reductase B/BLVRB Protein has been validated for the following applications: Enzyme Activity.