Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:C. tetani
- Size:10 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
- Endotoxin content:<1.0 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:1061100
- Protein synonyms:Tetanus toxin chain L|Tetanus toxin heavy chain|TeNT-LC|Tetanus toxin chain H|tetX|Tetanus toxin light chain|Tentoxylysin
- Accession no.:P04958
- Protein/peptide name:TeNT Light Chain/Tetanus Toxin
- Purity:>85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain at 5 ╡g per lane
- Molecular weight:43-50 kDa, reducing conditions
- Cat. no.:102877-116
- Supplier No.:6535-ZN-010
The Recombinant Tetanus Neurotoxin Type Light Chain Protein from R&D Systems is derived from E. coli. The Recombinant Tetanus Neurotoxin Type Light Chain Protein has been validated for the following applications: Enzyme Activity.