Human Recombinant Protocadherin alpha 4 (from CHO Cells)
Catalog # 102877-102
Supplier: R&D Systems
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Size:50 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin content:<1.0 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:56144
- Protein synonyms:MGC142169|CNR1|MGC138307|PCDH-ALPHA4|PCDHA4|ortholog of mouse CNR1|protocadherin alpha 4|protocadherin alpha-4|KIAA0345-like 10|PCDH-alpha-4|CRNR1|CNRN1
- Accession no.:NP_113688
- Protein/peptide name:Protocadherin alpha 4
- Purity:>90%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular weight:80-100 kDa, reducing conditions
- Cat. no.:102877-102
- Supplier No.:6509-CA-050
Specifications
About this item
The Recombinant Human Protocadherin alpha 4 Protein from R&D Systems is derived from CHO. The Recombinant Human Protocadherin alpha 4 Protein has been validated for the following applications: Bioactivity.